missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Piperonylbutoxide, MilliporeSigma™ Supelco™

Catalog No. 111020032
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020032 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020032 Supplier Merck Emd Millipore Supplier No. 45626100MG
Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 51-03-6
Density 1.059 g/mL (at 25°C)
Flash Point 171°C
Physical Form Neat
Linear Formula C19H30O5
MDL Number MFCD00005842
Boiling Point 155°C (0.3 mmHg)
Synonym 2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether; 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether
Type Analytical Standard
Quantity 100 mg
Formula Weight 338.44
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH