missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Phosmet, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 732-11-6 |
| Physical Form | Neat |
| Linear Formula | C11H12 NO4 PS2 |
| MDL Number | MFCD00055501 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 317.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH