missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Phenol Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 108-95-2 |
| Flash Point | 2°C |
| Linear Formula | C6H6 O |
| MDL Number | MFCD00002143 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 94.11 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in acetonitrile |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH