missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Phenmedipham, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 13684-63-4 |
| Flash Point | 100°C - Closed Cup |
| Physical Form | Neat |
| Linear Formula | C16H16N2O4 |
| MDL Number | MFCD00055419 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 300.31 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH