missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Perthan, MilliporeSigma™ Supelco™

Catalog No. 111020642
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020642 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020642 Supplier Merck Emd Millipore Supplier No. 45615250MG
Agriculture

Specifications

Product Line PESTANAL
CAS 72-56-0
Flash Point >100°C
Physical Form Neat
Linear Formula C18H20 Cl2
MDL Number MFCD00045254
Type Analytical Standard
Quantity 250 mg
Formula Weight 307.26
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH