missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Permethrin, MilliporeSigma™ Supelco™

Catalog No. 111020245
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020245 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020245 Supplier Merck Emd Millipore Supplier No. 45614250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Permethrin belongs to the pyrethroid class of insecticides.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 52645-53-1
Flash Point 542°C
Physical Form Neat
Linear Formula C21H20 Cl2O3
MDL Number MFCD00041809
Type Analytical Standard
Quantity 250 mg
Formula Weight 391.29
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH