missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Penthiopyrad, MilliporeSigma™ Supelco™

Catalog No. 111018601
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018601 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018601 Supplier Merck Emd Millipore Supplier No. 3206425MG

Penthiopyrad is a carboxanilide fungicide, mostly used against basidiomycetes and ascomycetes.1

Agriculture

Specifications

Product Line PESTANAL
CAS 183675-82-3
Physical Form Neat
Linear Formula C16H20F3N3 OS
MDL Number MFCD11973641
Synonym (RS)-N-[2-(1,3-Dimethylbutyl)-3-thienyl]-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 359.41
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH