missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Penflufen, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 494793-67-8 |
| Physical Form | Neat |
| Linear Formula | C18H24 FN3 O |
| MDL Number | MFCD22200854 |
| Synonym | 2 ′-[(RS)-1,3-Dimethylbutyl]-5-fluoro-1,3-dimethylpyrazole-4-carboxanilide; N-[2-(1,3-Dimethylbutyl)phenyl]-5-fluoro-1,3-dimethyl-1 H-pyrazole-4-carboxamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 317.40 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH