missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Parathion Solution, 100 μg/mL in cycloHexane, MilliporeSigma™ Supelco™

Catalog No. 111020013
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020013 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020013 Supplier Merck Emd Millipore Supplier No. 458902ML
Only null left
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 56-38-2
Flash Point -18°C
Linear Formula C10H14 NO5 PS
MDL Number MFCD00036219
Synonym Parathion-ethyl solution
Type Analytical Standard
Quantity 2 mL
Formula Weight 291.26
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in cyclohexane
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH