missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Parathion-methyl, MilliporeSigma™ Supelco™

Catalog No. 111019734
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019734 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019734 Supplier Merck Emd Millipore Supplier No. 36187100MG
Only null left

Parathion-methyl is an organophosphorus insecticide and acaricide, which is widely used in controlling insect pests of agricultural crops, fruits and vegetables.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 298-00-0
Flash Point 30°C
Physical Form Neat
Linear Formula C8H10 NO5 PS
MDL Number MFCD00041813
Synonym O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate; Methyl parathion
Type Analytical Standard
Quantity 100 mg
Formula Weight 263.21
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH