missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Oxasulfuron, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 144651-06-9 |
| Physical Form | Neat |
| Linear Formula | C17H18N4O6 S |
| MDL Number | MFCD00871602 |
| Synonym | Oxetan-3-yl 2-[(4,6-dimethylpyrimidin-2-yl)carbamoylsulfamoyl]benzoate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 406.41 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH