missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Orthosulfamuron, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 213464-77-8 |
| Physical Form | Neat |
| Linear Formula | C16H20N6O6 S |
| MDL Number | MFCD09027340 |
| Synonym | 1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[2-(dimethylcarbamoyl)phenylsulfamoyl]urea |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 424.43 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH