missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Ofurace, MilliporeSigma™ Supelco™

Catalog No. 111014769
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014769 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014769 Supplier Merck Emd Millipore Supplier No. 46143100MGR
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 58810-48-3
Physical Form Neat
Linear Formula C14H16 ClNO3
MDL Number MFCD01632753
Type Analytical Standard
Quantity 100 mg
Formula Weight 281.73
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH