missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Nonylphenol, technical Mixture, MilliporeSigma™ Supelco™

Catalog No. 111020324
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020324 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020324 Supplier Merck Emd Millipore Supplier No. 460181G
Only null left

Nonylphenol belongs to the class of alkylphenols, commonly employed as a precursor for the commercially important nonylphenol polyethoxylate, a non-ionic surfactant that is widely used for a broad-spectrum of applications in commercial and household formulations.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 84852-15-3
Density 0.937 g/mL (at 25°C)
Flash Point 154°C
Physical Form Neat
Linear Formula C15H24 O
Type Analytical Standard
Quantity 1 g
Formula Weight 220.35
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH