missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Nitrothal-isopropyl, MilliporeSigma™ Supelco™

Catalog No. 111012379
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012379 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012379 Supplier Merck Emd Millipore Supplier No. 36179100MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 10552-74-6
Physical Form Neat
Linear Formula C14H17 NO6
MDL Number MFCD00055456
Synonym Diisopropyl 5-nitroisophthalate
Type Analytical Standard
Quantity 100 mg
Formula Weight 295.29
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH