missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Napropamid, MilliporeSigma™ Supelco™

Catalog No. 111013224
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013224 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013224 Supplier Merck Emd Millipore Supplier No. 36175100MG
Agriculture

Specifications

Product Line PESTANAL
CAS 15299-99-7
Physical Form Neat
Linear Formula C10H7 OCH(CH3)CON(CH2 CH3)2
MDL Number MFCD00055354
Melting Point 73°C to 78°C
Synonym N,N-Diethyl-2-(1-naphthyloxy)propanamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 271.35
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH