missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Monocrotophos, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 6923-22-4 |
| Physical Form | Neat |
| Linear Formula | C7H14 NO5 P |
| MDL Number | MFCD00055291 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 223.16 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH