missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Metribuzin, MilliporeSigma™ Supelco™

Catalog No. 111020326
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020326 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020326 Supplier Merck Emd Millipore Supplier No. 36165100MG
Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 21087-64-9
Physical Form Neat
Linear Formula C8H14N4 OS
MDL Number MFCD00055525
Type Analytical Standard
Quantity 100 mg
Formula Weight 214.29
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH