missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Metolachlor, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 51218-45-2 |
| Physical Form | Neat |
| Linear Formula | C15H22 ClNO2 |
| MDL Number | MFCD00055293 |
| Synonym | 2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(-2-methoxy-1-methylethyl)acetamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 283.79 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH