missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Metolachlor ESA sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111015952
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015952 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015952 Supplier Merck Emd Millipore Supplier No. 3414910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 947601-85-6
Physical Form Neat
Linear Formula C15H22 NNaO5 S
MDL Number MFCD08702644
Synonym 2-[(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)-amino]2-oxo-ethanesulfonic acid sodium salt
Type Analytical Standard
Quantity 10 mg
Formula Weight 351.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH