missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Methomyl, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 16752-77-5 |
| Physical Form | Neat |
| Linear Formula | C5H10N2O2 S |
| MDL Number | MFCD00055451 |
| Melting Point | 77°C to 82°C |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 162.21 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH