missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Methiocarb sulfone, MilliporeSigma™ Supelco™

Catalog No. 111013556
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013556 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013556 Supplier Merck Emd Millipore Supplier No. 45729100MG
Agriculture

Specifications

Product Line PESTANAL
CAS 2179-25-1
Physical Form Neat
Linear Formula C11H15 NO4 S
MDL Number MFCD00144741
Synonym Mercaptodimethursulfon; Methiocarb sulfone
Type Analytical Standard
Quantity 100 mg
Formula Weight 257.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH