missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Methiocarb, MilliporeSigma™ Supelco™

Catalog No. 111020134
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020134 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020134 Supplier Merck Emd Millipore Supplier No. 36152100MG
Only null left

Methiocarb is a broad spectrum N-methylcarbamate pesticide widely used to control snails in orchids and is also a chemical bird repellent in fruit crops.

Agriculture

Specifications

Product Line PESTANAL
CAS 2032-65-7
Physical Form Neat
Linear Formula C11H15 NO2 S
MDL Number MFCD00055450
Synonym Mercaptodimethur
Type Analytical Standard
Quantity 100 mg
Formula Weight 225.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH