missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Methamidophos, MilliporeSigma™ Supelco™

Catalog No. 111019711
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019711 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019711 Supplier Merck Emd Millipore Supplier No. 33395100MG
Only null left

Methamidophos is a broad spectrum organophosphate pesticide used for controlling a variety of plant and soil pests in agricultural crops.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 10265-92-6
Flash Point 212°C - Closed Cup
Physical Form Neat
Linear Formula C2H8 NO2 PS
MDL Number MFCD00041808
Melting Point 42°C to 45°C
Synonym O,S-Dimethyl phosphoramidothioate
Type Analytical Standard
Quantity 100 mg
Formula Weight 141.13
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH