missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Methacrifos, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 62610-77-9 |
| Physical Form | Neat |
| Linear Formula | C7H13O5 PS |
| MDL Number | MFCD16661253 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 240.21 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH