missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Metaldehyde, MilliporeSigma™ Supelco™

Catalog No. 111019306
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019306 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019306 Supplier Merck Emd Millipore Supplier No. 366111GR
Only null left

Metaldehyde is an active ingredient, commonly used in organic pesticides, which can be widely used in agriculture in order to control mollusca species like snails, slug etc.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 9002-91-9
Physical Form Neat
Linear Formula (CH3 CHO)n
MDL Number MFCD00134115
Melting Point 190°C to 196°C (sublimation)
Synonym Acetaldehyde polymerized
Type Analytical Standard
Quantity 1 g
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH