missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Metalaxyl-(phenyl-13C6), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013001
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013001 Supplier Merck Emd Millipore Supplier No. 122881MG
Only null left

Specifications

Product Line PESTANAL
CAS 1356199-69-3
Flash Point 100°C
Linear Formula 13C6C9H21 NO4
MDL Number MFCD19687017
Melting Point 69°C to 73°C
Type Analytical Standard
Quantity 1 mg
Formula Weight 285.29
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH