missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Metaflumizone, MilliporeSigma™ Supelco™

Catalog No. 111018711
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018711 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018711 Supplier Merck Emd Millipore Supplier No. 32966100MG
Agriculture

Specifications

Product Line PESTANAL
CAS 139968-49-3
Physical Form Neat
Linear Formula C24H16F6N4O2
MDL Number MFCD08690508
Synonym 2 ′-[2-(4-Cyanophenyl)-1-(alpha,alpha,alpha-trifluoro-m-tolyl)ethylidene]-4-(trifluoromethoxy)carbanilohydrazide
Type Analytical Standard
Quantity 100 mg
Formula Weight 506.40
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH