missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Mepiquat-d16 chloride, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | PESTANAL |
| Linear Formula | C7D16ClN |
| Synonym | 1,1-Dimethylpiperidinium-d16 chloride |
| Quantity | 5 mg |
| Formula Weight | 165.76 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH