missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Mepiquat chloride, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 24307-26-4 |
| Physical Form | Neat |
| Linear Formula | C7H16 ClN |
| MDL Number | MFCD00055543 |
| Synonym | 1,1-Dimethylpiperidinium chloride |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 149.66 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH