missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Mefenpyr-diethyl, MilliporeSigma™ Supelco™

Catalog No. 111013000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013000 Supplier Merck Emd Millipore Supplier No. 46302100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mefenpyr-diethyl is a herbicide safener, used for post emergence control of selective grass weeds affecting crops such as wheat, rye and spring barley.

Agriculture

Specifications

Product Line PESTANAL
CAS 135590-91-9
Physical Form Neat
Linear Formula C16H18 Cl2N2O4
MDL Number MFCD09753375
Synonym 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1 H-pyrazole-3,5-dicarboxylic acid diethyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 373.23
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH