missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Mecoprop-P, MilliporeSigma™ Supelco™

Catalog No. 111020132
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020132 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020132 Supplier Merck Emd Millipore Supplier No. 36773250MGR

Mecoprop-P, a phenoxyalkanoic acid is widely used as herbicide. It forms colourless and odourless crystals. The (R)-isomer of mecoprop is the active herbicide site. It is generally used for postemergence control of broad-leaved weeds by translocating the herbicide to the roots.

Agriculture

Specifications

Product Line PESTANAL
CAS 16484-77-8
Physical Form Neat
Linear Formula C10H11 ClO3
MDL Number MFCD00144037
Synonym (R)-2-(4-Chloro-2-methylphenoxy)propionic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 214.65
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH