missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Mecarbam, MilliporeSigma™ Supelco™

Catalog No. 111020010
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020010 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020010 Supplier Merck Emd Millipore Supplier No. 36515100MG
Agriculture

Specifications

Product Line PESTANAL
CAS 2595-54-2
Flash Point >100°C
Physical Form Neat
Linear Formula C10H20 NO5 PS2
MDL Number MFCD00129669
Type Analytical Standard
Quantity 100 mg
Formula Weight 329.37
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH