missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Maneb, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 12427-38-2 |
| Physical Form | Neat |
| Linear Formula | [C4H6 MnN2S4 ]n |
| MDL Number | MFCD00046858 |
| Synonym | Manganese ethylenebis(dithiocarbamate) |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH