missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Mancozeb, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017275
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017275 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017275 Supplier Merck Emd Millipore Supplier No. 45553100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 8018-01-7
Product Line PESTANAL
Physical Form Neat
MDL Number MFCD00078616
Synonym Manganese Zinc ethylenebis(dithiocarbamate)
Quantity 100 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH