missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Isofenphos Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 25311-71-1 |
| Linear Formula | C15H24 NO4 PS |
| MDL Number | MFCD00041805 |
| Synonym | 2-(Ethoxy-isopropylamino-thiophosphoryloxy)benzoic acid isopropyl ester |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 345.39 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in acetonitrile |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH