missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Indaziflam, MilliporeSigma™ Supelco™

Catalog No. 111018266
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018266 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018266 Supplier Merck Emd Millipore Supplier No. 32096100MG

Indaziflam is a pre-emergent, alkylazine herbicide, used in weed control management. Its mode of action involves inhibiting cellulose wall synthesis in susceptible plants.

Agriculture

Specifications

Product Line PESTANAL
CAS 950782-86-2
Physical Form Neat
Linear Formula C16H20 FN5
MDL Number MFCD17167079
Synonym 2-Amino-4-[(1 R,2 S)-2,6-dimethyl-indan-1-ylamino]-6-(1-fluoroethyl)-1,3,5-triazine; N-[(1 R,2 S)-2,3-Dihydro-2,6-dimethyl-1 H-inden-1-yl]-6-[(1 RS)-1-fluoroethyl]-1,3,5-triazine-2,4-diamine
Type Analytical Standard
Quantity 100 mg
Formula Weight 301.36
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH