missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Imazapic, MilliporeSigma™ Supelco™

Catalog No. 111012359
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012359 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012359 Supplier Merck Emd Millipore Supplier No. 34179100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Imazapic is an imidazolinone herbicide which is effective against weedy rice constraints.

Agriculture

Specifications

Product Line PESTANAL
CAS 104098-48-8
Physical Form Neat
Linear Formula C14H17N3O3
MDL Number MFCD04307828
Synonym 2-(4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methylnicotinic acid; 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1 H-imidazol-2-yl]-5-methyl-3-pyridinecarboxylic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 275.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH