missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Hydramethylnon, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018532
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018532 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018532 Supplier Merck Emd Millipore Supplier No. 35373100MG
Agriculture

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line PESTANAL
CAS 67485-29-4
Physical Form Neat
Linear Formula C25H24F6N4
MDL Number MFCD00128045
Synonym 1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 2-(1,4,5,6-tetrahydro-5,5-dimethyl-2-pyrimidinyl)hydrazone; NSC 379665; Tetrahydro-5,5-dimethyl-2(1 H)-pyrimidinone[3-[4-(trifluoromethyl)phenyl]-1-[2-[4-(trifluoromethyl)phenyl]ethenyl]-2-propylidene]hydrazone
Type Analytical Standard
Quantity 100 mg
Formula Weight 494.48
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH