missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Hexazinone, MilliporeSigma™ Supelco™

Catalog No. 111019713
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019713 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019713 Supplier Merck Emd Millipore Supplier No. 36129100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hexazinone is a systemic triazine herbicide, which can find applications in weed control management, via inhibition of photosynthesis in the target plants.

Agriculture

Specifications

Product Line PESTANAL
CAS 51235-04-2
Physical Form Neat
Linear Formula C12H20N4O2
MDL Number MFCD00055526
Type Analytical Standard
Quantity 100 mg
Formula Weight 252.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH