missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Hexaconazol, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 79983-71-4 |
| Physical Form | Neat |
| Linear Formula | C14H17 Cl2N3 O |
| MDL Number | MFCD00144441 |
| Synonym | (+/-)-2-(2,4-Dichlorophenyl)-1-(1 H-1,2,4-triazol-1-yl)-2-hexanol |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 314.21 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH