missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Heptachlor, MilliporeSigma™ Supelco™

Catalog No. 111018527
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018527 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018527 Supplier Merck Emd Millipore Supplier No. 32349100MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 76-44-8
Physical Form Neat
Linear Formula C10H5 Cl7
MDL Number MFCD00213497
Synonym 1,4,5,6,7,8,8-Heptachloro-3 A,4,7,7 A-tetrahydro-4,7-methanoindene
Type Analytical Standard
Quantity 100 mg
Formula Weight 373.32
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH