missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Heptachlor endo-epoxide, isomer A, MilliporeSigma™ Supelco™

Catalog No. 111018419
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018419 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018419 Supplier Merck Emd Millipore Supplier No. 3549250MG
Only null left
Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 28044-83-9
Physical Form Neat
Linear Formula C10H5 Cl7 O
MDL Number MFCD00872082
Synonym endo-1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-4,7-methano-3 A,4,7,7 A-tetrahydroindane
Type Analytical Standard
Quantity 50 mg
Formula Weight 389.32
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH