missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ HCH, Mixture of isomers (:::=1:1:1:1), MilliporeSigma™ Supelco™

Catalog No. 111020135
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020135 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020135 Supplier Merck Emd Millipore Supplier No. 36756250MG
Only null left

Hexachlorocyclohexane (HCH) is an organochlorine pesticide with carcinogenic and mutagenic effects and also is a potential environmental contaminant.

Agriculture

Specifications

Product Line PESTANAL
CAS 608-73-1
Physical Form Neat
Linear Formula C6H6 Cl6
MDL Number MFCD00003825
Type Analytical Standard
Quantity 250 mg
Formula Weight 290.83
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH