missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Haloxyfop-P-methyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 72619-32-0 |
| Physical Form | Neat |
| Linear Formula | C16H13 ClF3 NO4 |
| MDL Number | MFCD04112762 |
| Synonym | (R)-2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridyloxy]phenoxy}propanoic acid methyl ester |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 375.73 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH