missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Glyphosate-2-13C, ≥99% (HPLC), MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 287399-31-9 |
| Linear Formula | HO2C13 CH2 NHCH2 PO(OH)2 |
| MDL Number | MFCD01075599 |
| Melting Point | 230°C |
| Synonym | N-(Phosphonomethyl)glycine-2-13 C |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 170.07 |
| Percent Purity | ≥99% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH