missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Glyphosate-2-13C,15N, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111017956
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017956 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017956 Supplier Merck Emd Millipore Supplier No. 904795MG
Only null left

Glyphosate is one of the most used herbicides, especially in agriculture applications such as weed and vegetation control.

Agriculture

Specifications

Product Line PESTANAL
CAS 285978-24-7
Linear Formula HO2C13 CH215 NHCH2 PO3H2
MDL Number MFCD01075407
Melting Point 230°C
Type Analytical Standard
Quantity 5 mg
Formula Weight 171.06
Percent Purity ≥99% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH