missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Gibberellic acid, 90-100% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015486
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015486 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015486 Supplier Merck Emd Millipore Supplier No. 36575250MG

Gibberellic acid is an endogenous plant growth regulator

Agriculture

Specifications

Product Line PESTANAL
CAS 77-06-5
Physical Form Neat
Linear Formula C19H22O6
MDL Number MFCD00079329
Synonym GA3; Gibberellin A3
Type Analytical Standard
Quantity 250 mg
Formula Weight 346.37
Percent Purity 90 to 100% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH