missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Furmecyclox, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 60568-05-0 |
| Physical Form | Neat |
| Linear Formula | C14H21 NO3 |
| MDL Number | MFCD00466943 |
| Synonym | N-Cyclohexyl-N-methoxy-2,5-dimethyl-3-furamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 251.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH