missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fosthiazate, MilliporeSigma™ Supelco™

Catalog No. 111018042
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018042 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018042 Supplier Merck Emd Millipore Supplier No. 3409950MG
Only null left

Fosthiazate is a non-fumigant, organophosphate nematicide, which shows broad-spectrum activity against a variety of plant parasitic nematodes, such as Meloidegyne spp., Globodera spp., and Pratylenchus spp.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 98886-44-3
Physical Form Neat
Linear Formula C9H18 NO3 PS2
MDL Number MFCD00274595
Synonym O-Ethyl S-(1-methylpropyl) 2-oxo-3-thiazolidinylphosphonothioate; S-sec-Butyl O-ethyl 2-oxo-3-thiazolidinylphosphonothioate
Type Analytical Standard
Quantity 50 mg
Formula Weight 283.35
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH